Would you like to log in again?
DiATOME® Cryo Immuno Diamond Knives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
DCIMM3530 DCIMM3520 DCIMM3530-R DCIMM3520-R
102681-512EA 3566.84 USD InStock
102681-512 102681-514 102681-510 102681-516
DiATOME® Cryo Immuno Diamond Knives, Electron Microscopy Sciences
Knives
The first cryo knife with a diamond platform, guarantees the best possible sectioning for cryo-immuno-chemistry.
The diamond platform guarantees easy and gentle section collection. Sections are collected directly from the diamond surface using a loop and a sucrose/methylcellulose droplet (Refs. Liou, Peters). This leads to a considerable reduction in mechanical stresses and therefore to improved structure preservation in sucrose-protected samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty