Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DiATOME® UltraTrim Trimming Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
UTT-R UWTT-40-R UWTT-R UTT-40-R
102681-530EA 1384.14 USD InStock
102681-530 102681-540 102681-532 102681-538
DiATOME® UltraTrim Trimming Tools, Electron Microscopy Sciences
Knives
Diatome Ultratrim allows quick, easy and accurate trimming at both room and cryo-temperatures.
Due to the extreme sharpness, less mechanical damage is applied to the sample during trimming.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty