Would you like to log in again?
mRNA Cap2'-O-Methyltransferase
Supplier: New England Biolabs
M0366S M0366S
102715-936EA 64 USD InStock
102715-936 CA102715-936
mRNA Cap2'-O-Methyltransferase
Enzymes
mRNA Cap 2´-O-Methyltransferase adds a methyl group at the 2´-O position of the first nucleotide adjacent to the cap structure at the 5´ end of the RNA
The cap-1 structure has been reported to enhance mRNA translation efficiency and hence may help improve expression in mRNA transfection and microinjection experiments. mRNA Cap 2´-O-Methyltransferase specifically requires RNA with an m7GpppN cap as substrate. It cannot utilize RNA with pN, ppN, pppN or GpppN at the 5´ end. Capped RNA may be prepared via in vitro transcription using cap analog or through enzymatic capping using the Vaccinia Capping Enzyme.
The enzyme utilizes S-adenosylmethionine (SAM) as a methyl donor to methylate capped RNA (cap-0) resulting in a cap-1 structure.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty