Would you like to log in again?
Instant Sticky-end Ligase Master Mix, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M0370L M0370S M0370L M0370S
102715-946EA 108 USD InStock
CA102715-944 102715-946 102715-944 CA102715-946
Instant Sticky-end Ligase Master Mix, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
Instant Sticky-end Ligase Master Mix is a ready-to-use 2X solution of T4 DNA ligase and a proprietary ligation enhancer in an optimized reaction buffer.
Specifically formulated to rapidly ligate cohesive-end (2–4 bp) substrates and improve transformation, the mix simplifies reaction set-up and provides an optimized ratio of enzyme and buffer components. No thawing of the master mix is required as it maintains a liquid state during storage at -20°C and no incubation time is necessary to achieve ligation efficiencies sufficient for successful sub-cloning of sticky-end substrates. Just add the master mix to DNA with compatible ends, mix and transform; thereby reducing the valuable time needed for routine ligations. Ligations for subcloning can be carried out in small volumes with low concentrations, allowing users to conserve precious DNA samples, and be used directly to transform many strains of chemically competent E. coli without dilution.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty