Would you like to log in again?
Galactose Oxidase (from Fusarium graminearum)
Supplier: Innovative Research
IFGGAOLY150UN IFGGAOLY450UN IFGGAOLY1KU
102770-944EA 242.22 USD InStock
102770-944 102770-940 102770-942
Galactose Oxidase (from Fusarium graminearum)
Enzymes
Animal free galactose oxidase purified lyophilized has been purified from Fusarium graminearum (previously misclassified as Dactylium dendroides). This is a lyophilized powder using sodium phosphate and sucrose as stabilizers.
Galactose oxidase is part of the oxioreductases family, and is a free radical enzyme that is unusually stable as compared to other protein free radicals. Galactose oxidase is useful for applications like bioanalysis (quanifying biomolecules using oxygen detection, colorimetric detection and cell labeling), organic synthesis (acting as a catalyst in synthesizing aldehydes and carboxylic acids from primary alcohols), and biometric compounds (developing models that mimic the structure and function of galactose oxidase), among others.
Caution: This material is sold for in vitro use only for manufacturing and research. This material is not suitable for human or animal use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty