Would you like to log in again?
QUANTIFOIL® Multi A Holey Carbon Films on Grids
Supplier: Electron Microscopy Sciences
Q325AMA Q210CMA Q210AMA Q410NMA Q225AMA Q310AMA Q450AMA Q425CMA Q4100AMA Q3100AMA Q250AMA Q2100AMA Q410AMA Q410CMA Q425AMA Q310NMA Q350AMA Q210NMA Q310CMA
76441-138PK 464.01 USD InStock
76441-138 76440-192 76441-234 76441-014 76441-068 76440-928 76440-608 76440-316 102773-230 76440-522 76440-830 76440-246 76440-554 76440-138 76440-732 76440-106 76440-960 76440-420 76440-662
QUANTIFOIL® Multi A Holey Carbon Films on Grids
Graticules Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) microscopy when compared with conventional holey film.
In the Multi A foil the pattern of hole sizes, shapes and arrangements is repeated. In addition to circular holes, the pattern includes oval-shaped ones which appear round at high tilt angles (~70°). The diameters of the round holes are about 1, 1.4, and 2 µm, and the bar widths range from 0.5 to 4 µm. The oval holes in the foil have a dimension of 8×2 µm and 4×1 µm.
The surface properties of QUANTIFOIL holey carbon support film, especially the wetting properties, may have to be adapted according to one's particular requirements. Untreated aging QUANTIFOIL® tends to be hydrophobic. Hydrophillicity of the foil can be achieved by glow discharging in residual air or by metal coating.
Ordering information: QUANTIFOIL® is delivered as a carbon foil; it can be reinforced with plastic film. The grids are packed with their coated side toward the center of the grid box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty