Would you like to log in again?
AP100 Grease, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60709
102773-574EA 373.16 USD InStock
102773-574
AP100 Grease, Electron Microscopy Sciences
Lubricants
Apiezon AP100 Grease is a silicone-free vacuum grease and lubricant. Containing PTFE, AP100 exhibits extremely high levels of lubricity and has been shown to provide eight times the level of lubrication offered by standard, petroleum-based lubricating greases. Apiezon AP100 is designed for use at ambient temperatures (generally between 10 to 30°C / 50 to 86°F).
  • Has anti-seize properties, making it an effective protector and lubricant in ultra-high vacuums
  • Exhibits extremely high levels of lubricity
  • Demonstrates an extremely low vapour pressure of 7 x 10-11 Torr at 20°C
  • Recommended for use at ambient temperatures
  • Does not suffer from contamination problems associated with silicone based greases such as "creep" or "carry over"
  • Easy to clean and remove using hot water and an aqueous glassware detergent, hydrocarbon or chlorinated solvents
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty