Would you like to log in again?
Wax, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60710 60713 60711 60712
102773-582EA 276.95 USD InStock
102773-582 102773-580 102773-576 102773-578
Wax, Electron Microscopy Sciences
Lubricants
The Apiezon range of waxes features three products including the popular Apiezon Wax W, also known as 'Black Wax'. All three waxes in the Apiezon range can be used as permanent or semi-permanent vacuum sealants or mounting media. Apiezon Wax W is also a perfect etch resist.
Apiezon Q Compound is an effective temporary vacuum sealant that is used as a short term solution in situations which do not allow for the immediate dismantling of a system for repair
Applications:
Apiezon Wax W: Hard Vacuum Sealing / Mounting Wax, Etch Resist
Apiezon Wax W100: Medium Hardness Vacuum Sealant / Mounting Wax
Apiezon Wax W40: Soft Vacuum Sealant / Mounting Wax
Apiezon Q Compound: Vacuum Sealing Compound
Apiezon Q Compound is an effective temporary vacuum sealant that is used as a short term solution in situations which do not allow for the immediate dismantling of a system for repair. It is a versatile product suitable for a multitude of uses, an excellent waterproof medium, and easy to use and to remove. Has 10 year shelf life.
It is a versatile product suitable for a multitude of uses, an excellent waterproof medium, and easy to use and to remove. Has 10 year shelf life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty