Would you like to log in again?
Combination Scales, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68073-PS300 68073-PS100 68073-PS150-UK 68073-PS300-UK 68073-PS150 68073-PS100-UK
102773-618EA 1540.21 USD InStock
102773-618 102773-620 102773-612 102773-610 102773-616 102773-614
Combination Scales, Electron Microscopy Sciences
Rulers
All of the PS-XO series of calibration slides is green float glass, bright chrome image.
PS300-XO: High definition 300 mm scale in 0.1 mm divisions with extended 0.5, 1.0 and 5.0 mm lines to allow calibrations in those increments too. Series of dots and crosses, at 10 mm intervals, included for calibrations where these shapes are preferred. Supplied in polished wooden case. Features 300 mm high definition scale in 0.1 mm divisions with additional dot and cross series at 10 mm centers.
PS150-XO: High definition 150 mm scale in 0.1 mm divisions with extended 0.5, 1.0, and 5.0 mm lines to allow calibrations in those increments too. Series of dots and crosses, at 10 mm intervals, included for calibrations where these shapes are preferred. Supplied in polished wooden case. Features 150 mm high definition scale in 0.1 mm divisions with additional dot and cross series at 10 mm centers.
PS100-XO: High definition 100 mm scale in 0.1 mm divisions with extended 0.5, 1.0, and 5.0 mm lines to allow calibrations in those increments too. Series of dots and crosses, at 10mm intervals, included for calibrations where these shapes are preferred. Supplied in polished wooden case. Features 100 mm high definition scale in 0.1 mm divisions with additional dot and cross series at 10 mm centers.
Series of dots and crosses, at 10 mm intervals, included for calibrations where these shapes are preferred. Supplied in polished wooden case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty