Would you like to log in again?
HIV PBMC, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PB009C-1-HIV PB009C-3-HIV PB009C-2-HIV
102830-126EA 1130.77 USD InStock
102830-126 102830-124 102830-128
HIV PBMC, Charles River Laboratories Cell Solutions
Blood
Human disease state peripheral blood mononuclear cells (PBMC) are collected from human volunteer donors under IRB approved informed consent by whole blood or leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty