Would you like to log in again?
Endothelial Lipase Human HEK293, 0.1 mg
Supplier: BioVendor
RD172173100
102830-614EA 583.35 USD InStock
102830-614
Endothelial Lipase Human HEK293, 0.1 mg
Enzymes
Total 490 AA. MW: 55.8 kDa (calculated). UniProtKB acc. No. Q9Y5X9 (Ser21–Pro500). N-terminal linker (2 extra AA), C-terminal linker (2 extra AA) and C-terminal His-tag (6 extra AA). Protein identity confirmed by LC-MS/MS.
  • Quality control test
  • BCA to determine quantity of the protein
  • SDS PAGE to determine purity of the protein
  • LAL to determine quantity of endotoxin
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty