Would you like to log in again?
NEBNEXT® ULTRA™ Directional RNA Second Strand Synthesis Module, New England Biolabs
Supplier: New England Biolabs
E7550L E7550S E7550S E7550L
102855-116EA 410 USD InStock
102855-116 102855-132 CA102855-116 CA102855-132
NEBNEXT® ULTRA™ Directional RNA Second Strand Synthesis Module, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The NEBNext® Ultra™ Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA
The dsDNA generated by the NEBNext Ultra Directional RNA Second Strand Module can be subsequently converted to blunt-end DNA fragments using the NEBNext Ultra End Prep Module.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty