Would you like to log in again?
XRN-1
Supplier: New England Biolabs
M0338L M0338S M0338S M0338L
102855-158EA 416 USD InStock
102855-158 CA102855-158 101449-366 CA101449-366
XRN-1
Enzymes
XRN-1 is highly processive 5´→3´ exoribonuclease, requiring 5´ monophosphate
It also acts on 5´ monophosphate ssDNA with greatly reduced efficiency. XRN-1 is not efficient in removing RNAs with recessed 5´P, like tRNA. It does not cleave dsDNA, ssDNA and RNA which contain di, triphosphate, OH and capped 5´ ends.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty