Would you like to log in again?
HISCRIBE™ T7 Quick High Yield RNA Synthesis Kit, New England Biolabs
Supplier: New England Biolabs
E2050S E2050S
102855-162EA 307 USD InStock
CA102855-162 102855-162
HISCRIBE™ T7 Quick High Yield RNA Synthesis Kit, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
The HiScribe T7 Quick High Yield RNA Synthesis Kit offers robust in vitro RNA transcription for a variety of applications
The master mix format allows for quick reaction setup. DNase I and LiCl are included for DNA template removal and quick RNA purification. In addition, the kit allows for capped RNA or dye labeled RNA synthesis by incorporation of cap analog (ARCA) or dye modified nucleotides
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty