Would you like to log in again?
Human Recombinant Carboxypeptidase B2
Supplier: Bon Opus Biosciences
CA30-10UG CA30-50UG CA30-500UG CA30-1MG
102863-628EA 497.54 USD InStock
102863-628 102863-624 102863-626 102863-622
Human Recombinant Carboxypeptidase B2
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human ribonuclease pancreatic is produced by our mammalian expression system and the target gene encoding Lys29-Thr156 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty