Would you like to log in again?
Human Recombinant Wbp2
Supplier: Bon Opus Biosciences
CE22-1MG CE22-50UG CE22-500UG CE22-10UG
102863-936EA 2487.69 USD InStock
102863-936 102863-938 102863-934 102863-940
Human Recombinant Wbp2
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human WW domain-binding protein 2 is produced by our E.coli expression system and the target gene encoding Met1-Ala100 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty