Would you like to log in again?
Human Recombinant Cryaa
Supplier: Bon Opus Biosciences
CF01-1MG CF01-10UG CF01-50UG CF01-500UG
102864-492EA 102.31 USD InStock
102864-492 102864-490 102864-488 102864-486
Human Recombinant Cryaa
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human alpha-crystallin A chain is produced by our E.coli expression system and the target gene encoding Met1-Ser173 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty