Would you like to log in again?
Dymon® LIQUID ALIVE® Odor Digester, Essendant
Supplier: Essendant
ITW33601
102865-820CS 103.29 USD InStock
102865-820
Dymon® LIQUID ALIVE® Odor Digester, Essendant
Deodorants and Air Fresheners
Eradicate nasty smells
Unique, industrial-strength formula contains enzyme-producing bacteria that digest malodors caused by dampness, mildew, mold, food, urine, vomit, fecal and organic matter. Great for use in schools, hotels, nursing homes and restaurants. Safe and easy to use. Won't stain carpets or upholstery. Odor control. Liquid. Neutral scent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty