Would you like to log in again?
BioFX® 450 nm Liquid Nova-Stop Solution for TMB Microwell Substrates, Surmodics
Supplier: Surmodics
BioFX®
NSTP-0100-01
102867-006EA 20 USD InStock
102867-006
BioFX® 450 nm Liquid Nova-Stop Solution for TMB Microwell Substrates, Surmodics
Assays ELISAs
The stop solution provides minimial signal drift compared to sulfuric or hydrochloric acid. Absorbance is read in the yellow spectral range (450 nm) and can be read up to 3 hours after the TMB reaction has been stopped using the Nova-Stop Solution.
  • Room Temperature Storage
  • 4 Year Shelf Life
  • Safe for lab personnel and environment (non-corrosive to both skin and eyes as opposed to the alternative sulfuric acid)
  • Ready to use
A liquid stop solution used in end point ELISA applications where a TMB substrate is used as the detection mechanism.
Certifications: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty