Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NEB®Stable Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
NEB®
C3040I C3040H C3040H C3040I
102877-560EA 282 USD InStock
CA102877-560 102877-560 CA102877-612 102877-612
NEB®Stable Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
Chemically competent E. coli cells suitable for high efficiency transformation and isolation of plasmid clones containing repeat elements.
  • Ideal for isolation of plasmid clones containing repeat elements and unstable inserts
  • Useful for isolating and propagating retroviral/lentiviral clones
  • Carries the endA mutation (in contrast to Stbl3) so that isolated plasmids are free of EndoI
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty