Would you like to log in again?
T4 RNA Ligase 1 (ssRNA Ligase) (30,000 units/ml), New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M0437M M0437M
102877-596EA 300 USD InStock
102877-596 CA102877-596
T4 RNA Ligase 1 (ssRNA Ligase) (30,000 units/ml), New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
T4 RNA Ligase 1, High Concentration (M0437M), is a 3X higher concentration than our standard T4 RNA Ligase 1 (M0204S/L)
This product is packaged with a vial of 100 mM ATP (N0451A). The increased concentration of reaction components is helpful in lower volume reactions where reagent volumes are limited. T4 RNA Ligase 1 catalyzes the ligation of a 5´ phosphoryl-terminated nucleic acid donor to a 3´ hydroxyl-terminated nucleic acceptor through the formation of a 3´ → 5´ phosphodiester bond, with hydrolysis of ATP to AMP and PPi. Substrates include singlestranded RNA and DNA as well as dinucleoside pyrophosphates.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty