Would you like to log in again?
beta-N-Acetylglucosaminidase S
Supplier: New England Biolabs
P0744S P0744L P0744S P0744L
102877-626EA 153 USD InStock
102877-626 CA102877-626 102877-624 CA102877-624
beta-N-Acetylglucosaminidase S
Enzymes
β-N-Acetylglucosaminidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing β-N-Acetylglucosamine residues from oligosaccharides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty