Would you like to log in again?
Quantitative Ashless Filter Papers, Advantec MFS
Supplier: Advantec
N05A18.5CM N0315.0CM N0618.5CM N05B18.5CM N04A5.5CM N04A9.0CM N05C5.5CM N077.0CM N05A7.0CM N05C11.0CM N0718.5CM N0611.0CM N04A7.0CM N04A18.5CM N05A4.25CM N039.0CM N067.0CM N05A12.5CM N0318.5CM N065.5CM N04A11.0CM N0711.0CM N05B7.0CM N079.0CM N05C9.0CM N05A24.0CM N075.5CM N05B11.0CM N0612.5CM N05A9.0CM N0311.0CM N0312.5CM N069.0CM N035.5CM N04A12.5CM N037.0CM N05A11.0CM N05B15.0CM N05B9.0CM N05A15.0CM N05B5.5CM N04A15.0CM N05B12.5CM N05C4.25CM N05C7.0CM N0715.0CM N05C18.5CM N0712.5CM N05C15.0CM N04A4.25CM N0615.0CM N05C12.5CM N05A5.5CM
102877-648PK 30.07 USD InStock
102877-648 102877-646 101102-374 101102-250 101102-180 102877-680 102877-640 102877-684 102877-682 102877-688 102877-644 102877-686 102877-642 102877-720 102877-714 102877-638 101102-306 102877-712 102877-636 102877-718 102877-716 101102-304 101102-344 101102-190 102877-670 102877-674 102877-672 102877-710 102877-678 102877-676 102877-704 102877-668 102877-702 102877-708 102877-706 101102-232 101102-432 102877-662 102877-660 102877-666 102877-700 102877-664 102877-658 101102-248 102877-692 102877-690 102877-696 102877-652 102877-694 102877-650 102877-656 102877-654 102877-698
Quantitative Ashless Filter Papers, Advantec MFS
Filters Filter Paper Quantitative Filter Paper
Advantec Quantitative Ashless Filter Papers are made from the highest quality alpha cotton cellulose.
Grade No.3 Medium retention, fast flow rate paper with good chemical resistance and wet strength. Recommended for analysis of soils, fertilizers, cement, and minerals. Grade No.5A Retain coarse and gelatinous ppt with high flow rates. Filter hydroxides, metalic aerosols, environmetnal monitoring, and determine silica content in steel. Grade No.5B Retain medium size ppt such as CaCo₃, PbSO₄, AgCl. General purpose paper. Grade No.5C High retention efficiency collecting fine ppt with slow flow rate. Gravimetric analysis and collecting particles that tend toward colloidal dispersion. Grade No.6 Retains medium-fine particulates. Uses are precision analysis of trace and precious metals, beverages, water. Grade No.7 Highest purity paper for medium particles. Fast flow rate. Recommended for precise gravimetric analysis of minute trace and precious metals. Grade No.4A Hardened, smooth low ash paper with superior wet strength. Retains fine crystalline ppt with slow flow rate. Suitable for Buchner filtrations, high pressure, high chemical and pH resistance.
One critical step in the manufacture of these filter papers is acid washing. All grades are double acid washed in hydrochloric then hydrofluoric acid to reduce levels of SiO₂, CaO, and Fe2O₃. Grade No.4A Hardened is further treated with nitric acid. Following the acid wash, all papers are rinsed in ultrapure water to neutralize. Ash Content: 0.01% (except No.4A 0.025%).
Ordering information: Various sizes up to 50.0 cm available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty