Would you like to log in again?
Clorox® Urine Remover
Supplier: Essendant
Clorox®
CLO31036CT
102877-764CS 132.86 USD InStock
102877-764
Clorox® Urine Remover
Cleaning Agents Detergents and Surface Cleansers
Clorox® Urine remover for stains and odors tackles the toughest urine stains and odors. With the cleaning power of hydrogen peroxide, it quickly breaks down urine to eliminate odors and remove stains. It removes bacteria, body fluids, germs, organic matter, pet stains or water-based soil.
  • Bleach-free
  • Formulated for tough-to-clean porous surfaces, like grout
  • Effective on soft, absorbent surfaces like carpet and mattresses
  • Smart Tube® Technology guarantees you will spray every drop
Ideal for use in hotels, long-term care facilities, locker rooms, schools and high-traffic restrooms. Can be used on concrete, walls and flooring, laundry pre-treatment, carpets, ceramics, cement, fabric, fiberglass, glass, grout, laminates, metal, plaster, plastic or vinyl.
Chemical Compound: Hydrogen Peroxide
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty