Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
NEBUILDER HiFi DNA Assembly® Master Mix, New England Biolabs
Supplier: New England Biolabs
E2621L E2621L E2621L E2621X E2621X E2621S E2621S E2621S E2621X
102877-802EA 683 USD InStock
102877-802 CA102877-802 FPE2621L CA102877-804 102877-806 CA102877-806 102877-804 FPE2621X FPE2621S
NEBUILDER HiFi DNA Assembly® Master Mix, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
NEBuilder HiFi DNA Assembly Master Mix was developed to improve the efficiency and accuracy of DNA assembly.
This method allows for seamless assembly of multiple DNA fragments, regardless of fragment length or end compatibility. This method has been used to assemble either single-stranded oligonucleotides or different sizes of DNA fragments with varied overlaps (15–80 bp). It has utility for the synthetic biology community, as well as those interested in one-step cloning of multiple fragments due to its ease of use, flexibility and simple master-mix format. The reaction includes different enzymes that work together in the same buffer. The exonuclease creates single-stranded 3´ overhangs that facilitate the annealing of fragments that share complementarity at one end (the overlap region). The polymerase fills in gaps within each annealed fragment. The DNA ligase seals nicks in the assembled DNA. The end result is a double-stranded fully sealed DNA molecule that can serve as template for PCR, RCA or a variety of other molecular biology applications, including direct transformation of E coli.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty