Would you like to log in again?
Rapid PNGase F
Supplier: New England Biolabs
P0710S P0711S P0710S P0711S
103218-942EA 516 USD InStock
103218-942 CA103218-942 102877-828 CA102877-828
Rapid PNGase F
Enzymes
Rapid PNGase F is an improved reagent that allows the complete and rapid deglycosylation of antibodies and fusion proteins in minutes
  • Non-reducing format
All N-glycans are released rapidly and without bias, and are ready to be prepared for downstream chromatography or mass spectrometry analysis. Rapid PNGase F creates an optimized workflow which reduces processing time without compromising sensitivity or reproducibility.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty