Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMB9 Mouse Monoclonal Antibody [clone: 792520]

Supplier: R & D Systems
MAB7709
102887-320EA 620.4 USD InStock
102887-320
Anti-PSMB9 Mouse Monoclonal Antibody [clone: 792520]
Antibodies
Type: Primary
Antigen: PSMB9
Clonality: Monoclonal
Clone: 792520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty