Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fellowes® Saturn™3i Laminators, Essendant

Supplier: Essendant
Fellowes®
FEL5736601
102902-512EA 268.57 USD InStock
102902-512
Fellowes® Saturn™3i Laminators, Essendant
Laminators
Laminating machine with a 12.5" wide entry and multiple temperature settings accommodates multiple document sizes.

  • Rapid one-minute warm-up with InstaHeat Technology
  • Compatible with 3- and 5-mil hot-lamination pouches as well as self-adhesive cold-lamination pouches
  • Green light and audible beep indicate when machine is ready to laminate
  • Release lever disengages pouch for re-centering or removal
  • Entry width accommodates multiple document sizes
  • Auto Shut-Off prevents overheating when not in use

Document size: 12" max.
Document thickness: 3 to 5 mil
Warm-up time: 1 minute
Laminating speed: 12" per minute

Ordering information: Includes ImageLast™ starter kit with ten 3-mil letter-sized pouches.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty