Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FN3K Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-13189R-CY3
10291-444EA 811.1 USD InStock
10291-444
Anti-FN3K Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Amines, including those present on proteins, spontaneously react with glucose to make fructosamines in a reaction termed glycation. Fructosamine 3-kinase (FN3K), a 309-amino acid enzyme initially identified in erythrocytes, catalyzes the ATP-dependent phosphorylation of the third carbon on both D- and L-fructosamines, leading to their destabilization and eventually, their removal from the protein. FN3K is a monomer that is ubiquitously expressed in mammalian tissue and phosphorylates both low molecular mass and protein-bound fructosamines which are formed as a result of glycation of glucose with primary amines. FN3K protects proteins from the harmful effects of nonenzymatic glycation, and may also be involved in peptide repair and cell metabolism. FN3KRP (fructosamine-3-kinase-related protein) is a 309 amino acid protein that is expressed in erythrocytes, bone marrow, spleen, brain and kidney and belongs to the fructosamine kinase family. FN3KRP functions to phosphorylate psicoamines and ribulosamines on the third carbon of their sugar moiety, thereby leading to the deglycation of the target amines.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FN3K
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty