Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fellowes® Spectra Laminator, Essendant

Supplier: Essendant
Fellowes®
FEL5738201
102938-046EA 100.86 USD InStock
102938-046
Fellowes® Spectra Laminator, Essendant
Laminators
User-friendly laminator for the home or home office features advanced temperature control for consistent results.

  • Compatible with 3- and 5-mil hot-lamination pouches as well as self-adhesive cold-lamination pouches
  • Release lever disengages pouch for re-centering or removal
  • Warms-up in just four minutes
  • Entry width accommodates multiple document sizes
  • Auto Shut-Off prevents overheating when not in use
  • Includes laminating starter kit

Document size: 9" max.
Document thickness: 3 to 5 mil
Warm-up time: 4 minutes
Laminating speed: 12" per minute

Ordering information: Includes ImageLast™ starter kit with ten 3-mil letter-sized pouches.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty