Would you like to log in again?
Human Recombinant Anthrax Toxin Receptor 1
Supplier: Bon Opus Biosciences
CA34-500UG CA34-1MG CA34-10UG CA34-50UG
102943-482EA 118.46 USD InStock
102943-482 102943-484 102943-486 102943-488
Human Recombinant Anthrax Toxin Receptor 1
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human anthrax toxin receptor 1 is produced by our mammalian expression system and the target gene encoding Glu33-Lys321 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty