Would you like to log in again?
Mouse Recombinant Sialic Acid Binding Ig-like Lectin 3
Supplier: Bon Opus Biosciences
CJ10-10UG CJ10-1MG CJ10-50UG CJ10-500UG
102944-288EA 2487.69 USD InStock
102944-288 102944-286 102944-284 102944-282
Mouse Recombinant Sialic Acid Binding Ig-like Lectin 3
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Sialic Acid Binding Ig-like Lectin 3 is produced by our Mammalian expression system and the target gene encoding Asp18-Glu240 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty