Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lipopolysaccharides (LPS) and Related Products, List Biological Laboratories
Supplier: List Biological Laboratories
201 423 304 314 401 301 433 421 225 434
102946-484EA 400 USD InStock
102946-484 102946-572 102946-582 102946-570 102946-492 102946-580 102946-576 102946-568 102946-578 102946-490
Lipopolysaccharides (LPS) and Related Products, List Biological Laboratories
Microbiology Test Systems Microorganism Tests
Bacterial lipopolysaccharides have long been recognized as the active component of gram negative bacterial endotoxins
LPS from Escherichia coli O111:B4 is a smooth strain of LPS that is isolated by a modification of the phenol extraction method of Westphal and Jann. Smooth strain LPS are dispersable in aqueous solvents at concentrations of up to 5.0 mg/ml.
These unique macromolecules have been extensively studied by investigators in many disciplines in efforts to elucidate and define relevant pathophysiologival parameters of endotoxin shock, a profound life-threatening consequence of bacterial sepsis. Lipopolysaccharides have generated intense interest as being among the most potent natural products capable of pluripotential immunostimulation manifested by the activation of host cells (e.g. B. lymphocytes, macrophages) to functional differentiation. Host cell activation by lipopolysaccharides produces a spectrum of hormone-active lymphokines and monokines, including interferons (alpha, beta and gamma), interleukins 1 and 6, tumor necrosis factor, platelet activating factor and procoagulant tissue factor. The documented capacity of lipopolysaccharids or their active lipid A component to initiate a variety of biochemical pathways (protein kinase C, cAMP dependent protein kinase, phosphatidyl inositol turnover, arachidonate metabolism, protein myristolation and activation of G-proteins) provides investigators with powerful molecular tools by which to study cellular activation mechanisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty