Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GOLGB1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-13356R-CY7
10296-584EA 811.1 USD InStock
10296-584
Anti-GOLGB1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
GM130, a cis-Golgi matrix protein, interacts specifically with p115 and provides a membrane docking site. Both GM130 and p115 are involved in vesicle tethering to Golgi membranes. The protein p115 also binds p400, alternatively called giantin. Giantin, the majority of whose mass projects into the cytoplasm, is involved in the docking of COPI vesicles via p115 to the Golgi membrane. Giantin, which also is known as macrogolgin or Golgi complex-associated protein, is involved in cross-bridge formation in the Golgi complex. Giantin, which can form a homodimer, is a single-pass type I membrane protein that is an antigen in Sjoegren syndrome and in chronic rheumatoid arthritis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Giantin/GM130
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty