Would you like to log in again?
CHROMagar™ KPC CHROMagar Base with KPC Supplement, Clinical Microbiology, Northeast Lab Services
Supplier: Northeast Lab Services
CHRP0001
102963-564PK 83.83 USD InStock
102963-564
CHROMagar™ KPC CHROMagar Base with KPC Supplement, Clinical Microbiology, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
CHROMagar Base is a unique culture medium for the direct isolation of bacteria harboring reduced susceptibility to carbapenems.
  • Detection after Overnight Incubation
  • Short Incubation
  • No Selective Pre-Enrichment Medium Needed
  • Direct Plating Option
A selective pre-enrichment medium is not needed, users can plate the sample directly, and incubation lasts only 18 to 24 hours.
The KPC Supplement has a shelf life of around two years for flexibility of use.
Caution: This item is for research use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty