Would you like to log in again?
β-Galactosidase Staining Kit, Cell Biolabs
Supplier: Cell Biolabs
AKR-100
102963-606EA 443.08 USD InStock
102963-606
β-Galactosidase Staining Kit, Cell Biolabs
Assays Cellular Assays
The Beta-Galactosidase Assay Kit uses a simple staining protocol to measure the transfection efficiency of the LacZ gene
The ß-Gal catalyzes the hydrolysis of X-Gal, which produces a blue color in cells expressing the gene. The kit provides sufficient reagents for 75 assays in 35 mm dishes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty