Would you like to log in again?
GFP ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
AKR-121-T AKR-121 AKR-121-5
102963-624EA 3686.15 USD InStock
102963-624 102963-622 102963-626
GFP ELISA Kit, Cell Biolabs
Assays ELISAs
Cell Biolabs’ GFP ELISA Kit is an enzyme immunoassay developed for detection and quantitation of GFP or GFP fusion protein in cell or tissue samples
The quantity of GFP or its variants (including BFP, CFP and YFP) in an unknown sample is determined by comparing its absorbance with that of a known recombinant GFP standard curve. The kit has detection sensitivity limit of 30 pg/mL GFP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty