Would you like to log in again?
His-Tag Protein ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
AKR-130
102963-630EA 683.08 USD InStock
102963-630
His-Tag Protein ELISA Kit, Cell Biolabs
Assays ELISAs
Cell Biolabs' His-Tag Protein ELISA Kit allows you to detect and quantify His-tagged protein samples simply and reliably by comparing your unknown samples to a known recombinant standard
The kit is compatible with proteins from cells or tissues, and works regardless of whether the His-tag is on the C-terminus or N-terminus. Sensitivity range of the kit is 4 µg/mL to 1 ng/mL, or 950 nM to 50 pM of 6xHis-tag residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty