Would you like to log in again?
RIPA Buffer, Cell Biolabs
Supplier: Cell Biolabs
AKR-190 AKR-191
102963-646EA 104.62 USD InStock
102963-646 102963-644
RIPA Buffer, Cell Biolabs
Cell Biolabs' RIPA Buffer is suitable for the extraction of cytoplasmic, membrane, and nuclear proteins from cells or tissues
The buffer is suitable for a variety of downstream applications such as reporter assays, protein assays, immunoassays and protein purification. RIPA Buffer is available with or without a separate Protease Inhibitor Cocktail.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty