Would you like to log in again?
Anoikis Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
CBA-080 CBA-081
102963-702EA 676.92 USD InStock
102963-702 102963-700
Anoikis Assay Kits, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Anoikis Assays allow you to quantify and monitor anchorage-dependent cell death in cells using a precoated plate
Live cells can be viewed under a microscope and quantified on a plate reader by MTT (colorimetric) or Calcein AM (fluorometric); both reagents are included in the kit. Dead cells are detected with the red EthD-1 reagent, which is also included. Assays are available in both 24-well and 96-well formats.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty