Would you like to log in again?
CytoSelect™ Cell Invasion Assay Kits, Collagen Coated, Cell Biolabs
Supplier: Cell Biolabs
CBA-111-COL CBA-110-COL CBA-112-COL
102963-762EA 910.77 USD InStock
102963-762 102963-756 102963-748
CytoSelect™ Cell Invasion Assay Kits, Collagen Coated, Cell Biolabs
Assays Cellular Assays
CytoSelect™ Cell Invasion Assays are ideal for determining the invasive properties of cells
With the CytoSelect™ Cell Invasion Assays, no manual cell counting is required. Quantitation of cells is performed using a colorimetric or fluorometric dye in a 96-well plate reader. The 8 µm pore size is suitable for most cell types including epithelial cells, fibroblasts, and cancer cell lines. Kits are available in 24-well and 96-well formats.
These kits use a Boyden Chamber in which inserts containing a membrane with a defined pore size are nested in the wells of a cell culture plate. The top of the insert membrane is coated with a thin layer of collagen gel. Invasive cells are seeded above the membrane and must degrade the collagen layer before migrating toward a chemoattractant in the lower chamber.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty