Would you like to log in again?
CytoSelect™ 24-Well Wound Healing Assay Kit, Cell Biolabs, Inc
Supplier: Cell Biolabs
CBA-120-5 CBA-120 CBA-120-T
102963-768EA 3181.54 USD InStock
102963-768 102963-766 102963-770
CytoSelect™ 24-Well Wound Healing Assay Kit, Cell Biolabs, Inc
Assays Immunological Assays
CytoSelect™ 24-Well Wound Healing Assay measures cell migration across a wound field gap in vitro
This assay provides a much more accurate and consistent method compared to the traditional scratch assay. Proprietary inserts generate a consistent 0.9mm wound gap between the cells. Cells may then be treated or monitored for proliferation or migration across the wound field by imaging samples at fixed time points or by time-lapse microscopy. The inserts are inert and leave no residues that could impede proliferation or migration of cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty