Would you like to log in again?
CytoSelect™ 96-well Cell Transformation Assays, Cell Biolabs
Supplier: Cell Biolabs
CBA-130 CBA-130-5 CBA-130-T
102963-796EA 393.85 USD InStock
102963-796 102963-794 102963-792
CytoSelect™ 96-well Cell Transformation Assays, Cell Biolabs
Assays Cellular Assays
CytoSelect™ 96-Well Cell Transformation Assay (Soft Agar Colony Formation) is suitable for measuring cell transformation where no downstream analysis is required
This assay uses a proprietary soft agar medium for superior results. Cells are incubated in a semisolid agar medium for 6-8 days, where they are allowed to form colonies. The medium is then solubilized, and the cells are lysed and detected in a fluorometric plate reader.
Total assay time is drastically reduced from 3 weeks to about 6-8 days, and no manual cell counting is required. Quantitation is attained using the provided fluorescent dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty