Would you like to log in again?
CytoSelect™ Cell Viability and Cytotoxicity Assay, Cell Biolabs
Supplier: Cell Biolabs
CBA-240
102963-842EA 393.85 USD InStock
102963-842
CytoSelect™ Cell Viability and Cytotoxicity Assay, Cell Biolabs
Assays Cellular Assays
The CytoSelect™ Cell Viability and Cytotoxicity Assay Kit provides a simple format for monitoring cell viability via metabolic activity
Prior to the assay, cells may be treated with compounds or agents that affect cell viability. This kit is suitable for eukaryotic cells, but not yeast or bacteria.
Live cells are detected with either MTT (colorimetric detection) or Calcein AM (fluorometric detection). Dead cells are detected by EthD-1 reagent (fluorometric). All 3 detection reagents are included, along with Saponin (a cell death initiator).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty