Would you like to log in again?
SNL Passage-Independent Feeder Cells, Cell Biolabs
Supplier: Cell Biolabs
CBA-316
102963-872EA 818.46 USD InStock
102963-872
SNL Passage-Independent Feeder Cells, Cell Biolabs
Cells
SNL 76/7 Feeder Cells are clonally derived from a mouse fibrobast STO cell line which has been transformed with neomycin resistance and murine LIF genes
This immortalized cell line has been shown to be useful as a feeder cell for ES cell growth as well as for culture of human and mouse iPS cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty