Would you like to log in again?
CytoSelect™ 96-Well Hematopoietic Colony Forming Cell Assay, Cell Biolabs
Supplier: Cell Biolabs
CBA-320 CBA-320-5
102963-874EA 566.15 USD InStock
102963-874 102963-876
CytoSelect™ 96-Well Hematopoietic Colony Forming Cell Assay, Cell Biolabs
Assays Cellular Assays
CytoSelect™ 96-Well Hematopoietic Colony Forming Cell Assay promotes the formation of HSC colonies in just 7-10 days
Methylcellulose medium supports growth of hematopoietic cell colonies. Full quantitation may be performed with no manual cell counting required. Cells can be either quantified in a fluorescence plate reader or recovered from the CFC assay medium for further downstream analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty