Would you like to log in again?
Cellular Senescence Detection Kit (SA-β-Gal Staining), Cell Biolabs
Supplier: Cell Biolabs
CBA-230-5 CBA-230
102964-644EA 553.85 USD InStock
102964-644 102964-646
Cellular Senescence Detection Kit (SA-β-Gal Staining), Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Cellular Senescence Staining Kit provides an efficient method to visualize Senescence Associated (SA) ß-galactosidase
SA-ß-Gal catalyzes the hydrolysis of X-gal, which produces a blue color in senescent cells. The assay is performed in standard 35 mm culture dishes, and results are visualized with a standard light microscope.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty