Would you like to log in again?
OxiSelect™ Nitrotyrosine ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-305 STA-305-5
102964-860EA 929.23 USD InStock
102964-860 102964-862
OxiSelect™ Nitrotyrosine ELISA Kit, Cell Biolabs
Assays ELISAs
OxiSelect™ Nitrotyrosine ELISA Kit provides a simple and sensitive method to measure the formation of 3-nitrotyrosine in proteins
Nitrotyrosine serves as a downstream marker for peroxynitrite and nitric oxide in a variety of disease states and in conditions of cellular damage and oxidative stress. Detect nitrotyrosine concentrations as low as 10 nM. The kit is suitable for use with cell lysates, serum, plasma and purified proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty