Would you like to log in again?
OxiSelect™ Protein Carbonyl ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
OxiSelect™
STA-310 STA-310-5 STA-310-T
102964-882EA 355 USD InStock
102964-882 102964-878 102964-880
OxiSelect™ Protein Carbonyl ELISA Kit, Cell Biolabs
Assays ELISAs
OxiSelect™ Protein Carbonyl ELISA Kit provides a rapid, efficient method for the detection of protein carbonyl residues
The carbonyl derivatives of proline, lysine, arginine and threonine residues are the most common products of protein oxidation in biological samples. Such derivatives are chemically stable and serve as markers for oxidative stress in most types of reactive oxygen species. The ELISA format is perfect for higher throughput and high sensitivity; concentration and precipitation steps have been eliminated, allowing greater sample retention.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty