Would you like to log in again?
OxiSelect™ AOPP Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
STA-318
102964-896EA 726.15 USD InStock
102964-896
OxiSelect™ AOPP Assay Kit, Cell Biolabs
Assays Enzyme Assays
OxiSelect™ AOPP Assay Kit provides a convenient method to assess oxidative stress
Advanced Oxidation Protein Products (AOPP) are uremic toxins created during oxidative stress through the reaction of plasma proteins with chlorinated oxidants such as chloramines or hypochlorous acid. AOPP levels are elevated in patients with renal complications, diabetes mellitus, and atherosclerosis, as well as HIV-positive patients.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty